Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloropentane, 98%, ACROS Organics™

Catalog No. p-90358
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle

CAS: 543-59-9 | C5H11Cl | 106.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-59-9
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.59
MDL Number MFCD00001015
InChI Key SQCZQTSHSZLZIQ-UHFFFAOYSA-N
Synonym pentyl chloride, pentane, 1-chloro, n-amyl chloride, n-pentyl chloride, amyl chloride, chloropentane, unii-0eg9msd3nk, 0eg9msd3nk, pentane, chloro, n-butylcarbonyl chloride
PubChem CID 10977
IUPAC Name 1-chloropentane
SMILES CCCCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -60°C
Color Colorless
Density 0.8800g/mL
Boiling Point 107°C to 108°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4115 to 1.4135
Linear Formula CH3(CH2)4Cl
Quantity 5 mL
Beilstein 01,13
Specific Gravity 0.88
Merck Index 15,6
Solubility Information Solubility in water: negligible. Other solubilities: miscible with alcohol and ether
Viscosity 0.59 mPa.s (20°C)
Formula Weight 106.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloropentane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-846-4
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.