Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorotridecane 98.0+%, TCI America™
SDP

Catalog No. C221325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C221325G 25 g
C22135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C221325G Supplier TCI America Supplier No. C221325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 822-13-9
Molecular Formula C13H27Cl
Molecular Weight (g/mol) 218.81
MDL Number MFCD03265416
InChI Key ASZMYJSJEOGSBR-UHFFFAOYSA-N
Synonym Tridecyl Chloride
PubChem CID 13193
IUPAC Name 1-chlorotridecane
SMILES CCCCCCCCCCCCCCl

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 g
UN Number 3082
Formula Weight 218.81
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorotridecane
EINECSNumber (2)-0066&(2)-0068
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.