Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexene-1-acetic acid

Catalog No. p-7049725
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 18294-87-6 | C8H12O2 | 140.182 g/mol

1-Cyclohexene-1-acetic acid used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18294-87-6
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00015482
InChI Key KDFBPHXESBPHTK-UHFFFAOYSA-N
Synonym 1-cyclohexenylacetic acid, 1-cyclohexene-1-acetic acid, 2-cyclohex-1-en-1-yl acetic acid, cyclohex-1-enylacetic acid, 1-cyclohexene-1-aceticacid, 2-cyclohexen-1-yl acetic acid, 2-cyclohex-1-enylacetic acid, cyclohex-1-en-1-ylacetic acid, 1-cyclohexenylacetate, cyclohexenylacetic acid
PubChem CID 86699
IUPAC Name 2-(cyclohexen-1-yl)acetic acid
SMILES C1CCC(=CC1)CC(=O)O

Specifications

Melting Point 33°C
Density 1.04 g/mL
Boiling Point 140°C to 142°C (25 mmHg)
Refractive Index 1.4775
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 140.18
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1-Cyclohexene-1-acetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 242-172-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.