Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexylpiperazine, 98%

Catalog No. AC134300050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 17766-28-8 | C10H20N2 | 168.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17766-28-8
Molecular Formula C10H20N2
Molecular Weight (g/mol) 168.28
MDL Number MFCD00044809
InChI Key XPDSXKIDJNKIQY-UHFFFAOYSA-N
Synonym n-cyclohexylpiperazine, 1-cyclohexyl-piperazine, cyclohexylpiperazine, piperazine, 1-cyclohexyl, 1-cyclohexylpiperazin, n-cyclohexyl piperazine, 1-cyclohexyl piperazine, piperazine,1-cyclohexyl, 4-1-cyclohexyl piperazine, acmc-1c47v
PubChem CID 87298
IUPAC Name 1-cyclohexylpiperazine
SMILES C1CCC(CC1)N2CCNCC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 28.0°C to 32.0°C
Color Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.6011
Quantity 5 g
Formula Weight 168.28
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 1-Cyclohexylpiperazine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
EINECSNumber 241-750-0
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.