Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrachloroethylene, 99%, extra pure
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Alkalinity | 0.003% max. (NaOH) |
| Melting Point | -22.0°C |
| Density | 1.6250g/mL |
| Boiling Point | 120.0°C to 122.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Plastic drum |
| Linear Formula | Cl2C=CCl2 |
| Refractive Index | 1.505 |
| Quantity | 10 L |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Suspected of causing cancer. Toxic to aquatic life with long lasting effects. Causes skin irritation. May cause an allergic skin reaction. May cause drowsiness or dizziness. Causes serious eye irritati |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at r |
| EINECSNumber | 204-825-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction