Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexylpiperidine 98.0+%, TCI America™
SDP

Catalog No. C303225ML
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 3319-01-5
Molecular Formula C11H21N
Molecular Weight (g/mol) 167.30
MDL Number MFCD00014642
InChI Key DSSKLTAHHALFRW-UHFFFAOYSA-N
Synonym cyclohexylpiperidine, n-cyclohexylpiperidine, piperidine, 1-cyclohexyl, 1-piperidinocyclohexane, 1-cyclohexyl-piperidine, acmc-1cpdp, cyclohexyl piperidine, wln: t6ntj a-al6tj, 5-20-02-00040 beilstein handbook reference
PubChem CID 18724
ChEBI CHEBI:38897
IUPAC Name 1-cyclohexylpiperidine
SMILES C1CCC(CC1)N2CCCCC2

Specifications

Color Yellow
Boiling Point 107°C
Quantity 25 mL
Formula Weight 167.30
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Cyclohexylpiperidine
RTECSNumber TM6520000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.