Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclopentenecarbonitrile, 95%

Catalog No. p-7028493
Click to view available options
Quantity:
1 g
5 g

CAS: 3047-38-9 | C6H7N | 93.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3047-38-9
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00043765
InChI Key XBVZRFXCDCYXAX-UHFFFAOYSA-N
Synonym 1-cyanocyclopentene, cyclopent-1-enecarbonitrile, 1-cyclopentenecarbonitrile, 1-cyclopentene-1-carbonitrile, cyclopent-1-ene-1-carbonitrile, 1-cyanocyclopenten, acmc-20aovl, 1-cyclopentenyl cyanide, 1-cyano-1-cyclopentene
PubChem CID 137808
IUPAC Name cyclopentene-1-carbonitrile
SMILES C1CC=C(C1)C#N

Specifications

Density 0.944
Boiling Point 72°C (15 mmHg)
Refractive Index 1.4715
Quantity 1 g
UN Number UN3276
Formula Weight 93.14
Percent Purity 97%
Chemical Name or Material 1-Cyclopentenecarbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.