Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%

Catalog No. AAH66672MD
Click to view available options
Quantity:
250 mg
1 g

CAS: 1151802-22-0 | C12H19BN2O2 | 234.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1151802-22-0
Molecular Formula C12H19BN2O2
Molecular Weight (g/mol) 234.106
MDL Number MFCD16659007
InChI Key NLWYVKHISUTBMY-UHFFFAOYSA-N
Synonym 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopropyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-di..., 1-cyclopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopropyl-4-pinacolylboronate-pyrazole, 1-cyclopropy-1-yl-boronic acid pinacol ester, 1-cyclopropyl 1h-pyrazole-4-boronic acid pinacol ester, 1h-pyrazole,1-cyclopropyl-4-4,4,5,5-tetramethyl-1, 1-cyclopropyl-1h-pyrazol-4-yl boronic acid pinacol ester
PubChem CID 59327133
IUPAC Name 1-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CC3

Specifications

Quantity 250 mg
Beilstein 19218005
Formula Weight 234.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.