1-Decene, 96%, remainder isomers

Manufacturer: Alfa Aesar 3121322

Quantity 100g

 View more versions of this product

Catalog No. AA3121322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 872-05-9
    Molecular Formula CH3(CH2)7CH=CH2
    Linear Formula CH3(CH2)7CH=CH2
    Formula Weight 140.27
    MDL Number MFCD00009577
    UN Number UN3295
    Beilstein 1737236
    Chemical Name or Material 1-Decene

    Specifications

    Formula Weight 140.27
    Quantity 100g
    TSCA TSCA
    Linear Formula CH3(CH2)7CH=CH2
    MDL Number MFCD00009577
    UN Number UN3295
    Beilstein 1737236
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H304-H226-H315
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    RTECS Number HE2071401
    Physical Form Liquid
    Density 0.741
    Assay Percent Range 96%
    Molecular Formula CH3(CH2)7CH=CH2
    CAS 872-05-9
    Chemical Name or Material 1-Decene
    Assay 96%, Remainder isomers

    GHS H Statement
    H304-H226-H315
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    Causes skin irritation.

    GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit