Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decyne, 98%

Catalog No. AAA1117114
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 764-93-2 | C10H18 | 138.254 g/mol

1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.

Solubility
Not miscible in water.

Notes
Incompatible with strong oxidizing agents. Store at 0-6°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 764-93-2
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00009576
InChI Key ILLHQJIJCRNRCJ-UHFFFAOYSA-N
Synonym 1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
PubChem CID 12997
ChEBI CHEBI:87322
IUPAC Name dec-1-yne
SMILES CCCCCCCCC#C

Specifications

Melting Point -44°C
Density 0.767
Boiling Point 174°C to 176°C
Flash Point 50°C (122°F)
Odor Petrol-like
Refractive Index 1.427
Quantity 25 g
UN Number UN3295
Beilstein 1236372
Solubility Information Not miscible in water.
Formula Weight 138.26
Percent Purity 98%
Chemical Name or Material 1-Decyne
Show More Show Less
Hazard Category H226-H315-H318
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 212-132-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.