Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Dimethylamino-2-propanol, 99%

Click to view available options
Quantity:
250g
1000g
108-16-7
C5H13NO
103.165
MFCD00004532
NCXUNZWLEYGQAH-UHFFFAOYSA-N
1-dimethylamino-2-propanol,dimepranol,1-dimethylamino propan-2-ol,n,n-dimethylisopropanolamine,dimethyl 2-hydroxypropyl amine,2-propanol, 1-dimethylamino,dimethylisopropanolamine,dimepranol inn,n,n-dimethylamino-2-propanol
37511
1-(dimethylamino)propan-2-ol
CC(CN(C)C)O
108-16-7
C5H13NO
103.165
MFCD00004532
NCXUNZWLEYGQAH-UHFFFAOYSA-N
1-dimethylamino-2-propanol,dimepranol,1-dimethylamino propan-2-ol,n,n-dimethylisopropanolamine,dimethyl 2-hydroxypropyl amine,2-propanol, 1-dimethylamino,dimethylisopropanolamine,dimepranol inn,n,n-dimethylamino-2-propanol
37511
1-(dimethylamino)propan-2-ol
CC(CN(C)C)O
Applications
1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv

Solubility
Fully miscible in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
pH 11
Boiling Point 123°C to 126°C
Quantity 250g
Odor Amine-like
Flash Point 35°C (95°F)
Melting Point -85°C
UN Number UN2734
Formula Weight 103.16
Percent Purity 99%
Chemical Name or Material 1-Dimethylamino-2-propanol

GHS H Statement
H314-H226-H302
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Harmful if swallowed.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

TSCA : TSCA