Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecanol, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187891
Click to view available options
Quantity:
1 mL
5 mL
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 500°F
CAS 112-53-8
Vapor Pressure 0.1 mmHg (20°C)
Vapor Density 7.4 (vs. air)
Density 0.833 g/mL (at 25°C)
Flash Point 121°C - Closed Cup
Physical Form Neat
Linear Formula CH3(CH2)11 OH
MDL Number MFCD00004753
Melting Point 22°C to 26°C
Boiling Point 260°C to 262°C
Synonym Alcohol C12; Dodecyl alcohol; Lauryl alcohol
Type Analytical Standard
Quantity 1 mL
Formula Weight 186.33
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.