Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecanol, 98%

Catalog No. AAA122280B
Click to view available options
Quantity:
250 g
1000 g
5000 g

CAS: 112-53-8 | C12H26O | 186.34 g/mol

1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.

Solubility
Soluble in water (0.004g/L).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00004753
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
Synonym 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
PubChem CID 8193
ChEBI CHEBI:28878
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

Melting Point 24°C to 27°C
Density 0.83
Boiling Point 258°C to 260°C
Flash Point 127°C (260°F)
Odor Alcohol-like
Assay Percent Range 98%
Refractive Index 1.443
Linear Formula CH3(CH2)11OH
Quantity 1000 g
UN Number UN3077
Beilstein 1738860
Merck Index 14,3405
Solubility Information Soluble in water (0.004g/L).
Formula Weight 186.34
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Dodecanol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-982-0
RTECSNumber JR5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.