Learn More
1-Dodecanol, 98%
Description
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.
Solubility
Soluble in water (0.004g/L).
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | 24°C to 27°C |
Density | 0.83 |
Boiling Point | 258°C to 260°C |
Flash Point | 127°C (260°F) |
Odor | Alcohol-like |
Assay Percent Range | 98% |
Refractive Index | 1.443 |
Linear Formula | CH3(CH2)11OH |
Quantity | 5000 g |
UN Number | UN3077 |
Show More |
Safety and Handling
Hazard Category | H315-H319 |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 203-982-0 |
RTECSNumber | JR5775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.