Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trimethylsilyl)acetamide, 95%

Catalog No. AC156461000
Click to view available options
Quantity:
25 mL
100 mL
1 L
Packaging:
Glass bottle

CAS: 10416-59-8 | C8H21NOSi2 | 203.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -8°C
Color Beige to Brown or Yellow
Density 0.8200g/mL
Boiling Point 71°C to 73°C (35.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4165 to 1.4185
Linear Formula CH3C[=NSi(CH3)3]OSi(CH3)3
Quantity 100 mL
Fieser 01,61; 02,30; 03,23; 13,34
Specific Gravity 0.82
Solubility Information Solubility in water: decomposes
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
Formula Weight 203.43
Percent Purity 95%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N, O-Bis(trimethylsilyl)acetamide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 233-892-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.