Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Dodecanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.
Solubility
Soluble in water (0.004g/L).
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.
Solubility
Soluble in water (0.004g/L).
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications
Melting Point | 24°C to 27°C |
Density | 0.830 |
Boiling Point | 258°C to 260°C |
Flash Point | 127°C (260°F) |
Assay Percent Range | 98% |
Refractive Index | 1.443 |
Linear Formula | CH3(CH2)11OH |
Quantity | 250 g |
UN Number | UN3077 |
Beilstein | 1738860 |
Show More |
Safety and Handling
Hazard Category | H315-H319 |
Hazard Statement | GHS H Statement H315-H319 |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 203-982-0 |
RTECSNumber | JR5775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction