Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecene, 93-95%

Catalog No. AC117630100
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
Plastic drum

CAS: 112-41-4 | C12H24 | 168.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-41-4
Molecular Formula C12H24
Molecular Weight (g/mol) 168.32
MDL Number MFCD00008961
InChI Key CRSBERNSMYQZNG-UHFFFAOYSA-N
Synonym 1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
PubChem CID 8183
IUPAC Name dodec-1-ene
SMILES CCCCCCCCCCC=C

Specifications

CAS Min % 93.0
CAS Max % 100.0
Melting Point -35.0°C
Density 0.7500g/mL
Boiling Point 213.0°C
Flash Point 77°C
Assay Percent Range 93% min. (GC)
Packaging Metal Drum
Refractive Index 1.4284 to 1.4304
Linear Formula CH3(CH2)9CH=CH2
Quantity 10 kg
Beilstein 01,225
Specific Gravity 0.75
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 168.32
Percent Purity 93 to 95%
Chemical Name or Material 1-Dodecene, 93-95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap an
EINECSNumber 203-968-4
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.