Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Eicosanethiol, 98%, Thermo Scientific Chemicals

Catalog Number AAH3449803
Click to view available options
Quantity:
250 mg
1 g
13373-97-2
C20H42S
314.616
MFCD07779406
YYHYWOPDNMFEAV-UHFFFAOYSA-N
1-eicosanethiol, eicosanethiol, 1-icosanethiol, icosyl mercaptan, eicosyl mercaptan, acmc-209btf
139443
icosane-1-thiol
CCCCCCCCCCCCCCCCCCCCS
13373-97-2
C20H42S
314.616
MFCD07779406
YYHYWOPDNMFEAV-UHFFFAOYSA-N
1-eicosanethiol, eicosanethiol, 1-icosanethiol, icosyl mercaptan, eicosyl mercaptan, acmc-209btf
139443
icosane-1-thiol
CCCCCCCCCCCCCCCCCCCCS

1-Eicosanethiol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Eicosanethiol is used as an organic chemical synthesis intermediate.

Notes
Air and moisture sensitive. Incompatible with heat.
TRUSTED_SUSTAINABILITY
Melting Point 38°C to 41°C
Odor Stench
Linear Formula CH3(CH2)19SH
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 314.61
Percent Purity 98%
Chemical Name or Material 1-Eicosanethiol
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit