Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Eicosanethiol, 98%

Catalog No. p-7032857
Click to view available options
Quantity:
250 mg
1 g

CAS: 13373-97-2 | C20H42S | 314.616 g/mol

1-Eicosanethiol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Eicosanethiol is used as an organic chemical synthesis intermediate.

Notes
Air and moisture sensitive. Incompatible with heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13373-97-2
Molecular Formula C20H42S
Molecular Weight (g/mol) 314.616
MDL Number MFCD07779406
InChI Key YYHYWOPDNMFEAV-UHFFFAOYSA-N
Synonym 1-eicosanethiol, eicosanethiol, 1-icosanethiol, icosyl mercaptan, eicosyl mercaptan, acmc-209btf
PubChem CID 139443
IUPAC Name icosane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCCCS

Specifications

Melting Point 38°C to 41°C
Odor Stench
Linear Formula CH3(CH2)19SH
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 314.61
Percent Purity 98%
Chemical Name or Material 1-Eicosanethiol
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.