Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Ethyl-2-methylquinolinium iodide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.
Solubility
Soluble in Water.
Notes
Precursor in synthesis of carbocyanine dyes.
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.
Solubility
Soluble in Water.
Notes
Precursor in synthesis of carbocyanine dyes.
Specifications
Melting Point | 231°C (decomposition) |
Quantity | 100 g |
Sensitivity | Light sensitive |
Solubility Information | Soluble in Water. |
Formula Weight | 299.16 |
Percent Purity | 97% |
Chemical Name or Material | 1-Ethyl-2-methylquinolinium iodide |
Safety and Handling
Hazard Category | H302+H312+H332 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 210-120-7 |
RTECSNumber | VA0740000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction