Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-2-pyrrolidinone, 98%

Catalog No. p-7027639
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 2687-91-4 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-91-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00003199
InChI Key ZFPGARUNNKGOBB-UHFFFAOYSA-N
Synonym 1-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, n-ethylpyrrolidinone, ethyl pyrrolidone, unii-h0229sx1cw, n-ethyl-, a-pyrrolidone, n-ethyl-2-pyrrolidinone
PubChem CID 17595
IUPAC Name 1-ethylpyrrolidin-2-one
SMILES CCN1CCCC1=O

Specifications

Density 0.992
Boiling Point 96°C to 98°C (20 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.465
Quantity 25 g
Formula Weight 113.16
Percent Purity 98%
Chemical Name or Material 1-Ethyl-2-pyrrolidinone
Hazard Category H227-H318-H360Df
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P281-P305+P351+P338-P308+P313-P310-P370+P378q-P501c
EINECSNumber 220-250-6
RTECSNumber UY5769250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.