Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-2-pyrrolidinone, 98%

Catalog No. AC163931000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163931000 100 g Glass bottle
AC163930250 25 g Glass bottle
AC163935000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC163931000 Supplier Thermo Scientific Chemicals Supplier No. 163931000
Add to Cart
This item is not returnable. View return policy

CAS: 2687-91-4 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-91-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00003199
InChI Key ZFPGARUNNKGOBB-UHFFFAOYSA-N
Synonym 1-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, n-ethylpyrrolidinone, ethyl pyrrolidone, unii-h0229sx1cw, n-ethyl-, a-pyrrolidone, n-ethyl-2-pyrrolidinone
PubChem CID 17595
IUPAC Name 1-ethylpyrrolidin-2-one
SMILES CCN1CCCC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -75.0°C
Density 0.9980g/mL
Boiling Point 212.5°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 100 g
Fieser 09,220
Specific Gravity 0.998
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Viscosity 2.1 mPa.s (20°C)
Formula Weight 113.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Ethyl-2-pyrrolidinone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 220-250-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.