Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium chloride, 97%

Catalog No. p-3736474
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

Catalog No. AC354080050

Supplier: Thermo Scientific Chemicals 354080050

Only null left
Add to Cart

CAS: 65039-09-0 | C6H11ClN2 | 146.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65039-09-0
Molecular Formula C6H11ClN2
Molecular Weight (g/mol) 146.62
MDL Number MFCD00074843
InChI Key BMQZYMYBQZGEEY-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics™ st 80, dsstox_cid_21180
PubChem CID 2734160
ChEBI CHEBI:61327
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;chloride
SMILES [Cl-].CCN1C=C[N+](C)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 77.0°C to 79.0°C
Color Yellow
Flash Point 186°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Viscosity 61.2 mPa.s (80°C)
Formula Weight 146.62
Percent Purity 97%
Physical Form Agglomerating Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.