Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%

Catalog No. AAL1976209
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1976209 10 g
AAL1976204 2 g
AAL1976218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1976209 Supplier Thermo Scientific Chemicals Supplier No. L1976209
Add to Cart

CAS: 155371-19-0 | C6H11F6N2P | 256.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155371-19-0
Molecular Formula C6H11F6N2P
Molecular Weight (g/mol) 256.132
MDL Number MFCD00216703
InChI Key DPDAKOVGQUGTHH-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc
PubChem CID 2758873
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate
SMILES CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 58°C to 62°C
Quantity 10 g
Sensitivity Hygroscopic
Formula Weight 256.13
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium hexafluorophosphate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.