Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium tetrafluoroborate, 98+% (dry wt.), may cont. up to 3% water

Catalog No. AAL1976309
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1976309 10 g
AAL1976304 2 g
AAL1976318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1976309 Supplier Thermo Scientific Chemicals Supplier No. L1976309
Add to Cart

CAS: 143314-16-3 | C6H11BF4N2 | 197.97 g/mol

1-Ethyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used in the recycling of osmium in the dihydroxylation of olefins with osmium(VIII)¬ oxide. It is a stable liquid electrolyte used in the capillary electrophoretic method for resolving phenolic compounds present in grape seed extract. It also acts as a good solvent for a wide range of both organic and inorganic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used in the recycling of osmium in the dihydroxylation of olefins with osmium(VIII)¬ oxide. It is a stable liquid electrolyte used in the capillary electrophoretic method for resolving phenolic compounds present in grape seed extract. It also acts as a good solvent for a wide range of both organic and inorganic compounds.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143314-16-3
Molecular Formula C6H11BF4N2
Molecular Weight (g/mol) 197.97
MDL Number MFCD00216668
InChI Key CUNYTRQQXKCRTJ-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium tetrafluoroborate, 1-ethyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate, emimbf4, basionics™ ee 03, acmc-209cq9, dsstox_cid_29074, dsstox_rid_83293, dsstox_gsid_49218, ksc174g8n, 1-methyl-3-ethylimidazolium tetrafluoroborate
PubChem CID 2769348
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCN1C=C[N+](C)=C1

Specifications

Melting Point 15°C
Density 1.28
Flash Point None
Refractive Index 1.41
Quantity 10 g
Sensitivity Hygroscopic
Formula Weight 197.98
Percent Purity ≥98%
Assay (dry wt.), may cont. up to 3% water
Chemical Name or Material 1-Ethyl-3-methylimidazolium tetrafluoroborate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.