Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-4-ethynylbenzene 97.0+%, TCI America™
SDP

Catalog No. E074925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E074925G 25 g
E07495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E074925G Supplier TCI America Supplier No. E074925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40307-11-7
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00173887
InChI Key ZNTJVJSUNSUMPP-UHFFFAOYSA-N
Synonym 4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
PubChem CID 142425
IUPAC Name 1-ethyl-4-ethynylbenzene
SMILES CCC1=CC=C(C=C1)C#C

Specifications

Color Yellow
Boiling Point 192°C
Quantity 25 g
Formula Weight 130.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-4-ethynylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.