Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-4-[(p-tolyl)ethynyl]benzene, 99+%, Thermo Scientific Chemicals

Catalog No. p-7029423
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22692-80-4
Molecular Formula C17H16
Molecular Weight (g/mol) 220.315
MDL Number MFCD04038793
InChI Key LFNSILZUJYWPJS-UHFFFAOYSA-N
Synonym 1-4-ethylphenyl-2-4-methylphenyl acetylene, 1-ethyl-4-p-tolyl ethynyl benzene, 1-ethyl-4-2-4-methylphenyl ethynyl benzene, 1-4-ethyl-4-p-tolyl ethylnyl benzene, acmc-20apau, 1-ethyl-4-p-tolylethynyl benzene, 1-4-ethylphenyl-2-p-tolyl acetylene, 1-ethyl-4-4-methylphenyl ethynyl benzene, benzene,1-ethyl-4-2-4-methylphenyl ethynyl, 1-ethyl-4-2-4-methylphenyl-1-ethynyl benzene
PubChem CID 11413368
IUPAC Name 1-ethyl-4-[2-(4-methylphenyl)ethynyl]benzene
SMILES CCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)C

Specifications

Melting Point 71°C to 74°C
Boiling Point 135°C to 136°C (1 mmHg)
Quantity 1 g
Formula Weight 220.31
Percent Purity ≥99%
Chemical Name or Material 1-Ethyl-4-[(p-tolyl)ethynyl]benzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.