Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-4-piperidone 98.0+%, TCI America™
SDP

Catalog No. E0498250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0498250ML 250 mL
E049825ML 25 mL
E0498100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0498250ML Supplier TCI America Supplier No. E0498250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3612-18-8
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00013356
InChI Key BDVKAMAALQXGLM-UHFFFAOYSA-N
Synonym 1-ethyl-4-piperidone, 4-piperidinone, 1-ethyl, n-ethyl-4-piperidone, pubchem6784, n-ethylpiperid-4-one, acmc-209ikz, 1-ethyl-4-piperidinone, 1-ethyl-4-oxopiperidine, 1-ethyl-piperidin-4-one, 1-ethyl piperidin-4-one
PubChem CID 77159
IUPAC Name 1-ethylpiperidin-4-one
SMILES CCN1CCC(=O)CC1

Specifications

Color Yellow
Boiling Point 80°C
Quantity 250 mL
Formula Weight 127.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-4-piperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.