Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethylcyclopentanol, 96%, Thermo Scientific Chemicals

Catalog No. AAH6680903
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1462-96-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00019281
InChI Key LPCWIFPJLFCXRS-UHFFFAOYSA-N
Synonym 1-ethylcyclopentanol, cyclopentanol, 1-ethyl, 1-ethyl-1-cyclopentanol, acmc-1bzot, ksc181q7f, 1-ethyl-1-hydroxycyclopentane
PubChem CID 137018
IUPAC Name 1-ethylcyclopentan-1-ol
SMILES CCC1(CCCC1)O

Specifications

Color Colorless to Yellow
Density 0.909 g/mL
Flash Point 49°C (120°F)
Refractive Index 1.457
Quantity 1 g
UN Number UN1987
Sensitivity Moisture sensitive
Formula Weight 114.19
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Ethylcyclopentanol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.