Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethylpyridinium bromide, 99%

Catalog No. p-7023898
Click to view available options
Quantity:
25 g
100 g

CAS: 1906-79-2 | C7H10BrN | 188.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1906-79-2
Molecular Formula C7H10BrN
Molecular Weight (g/mol) 188.068
MDL Number MFCD00041999
InChI Key ABFDKXBSQCTIKH-UHFFFAOYSA-M
Synonym 1-ethylpyridinium bromide, 1-ethylpyridin-1-ium bromide, ethylpyridinium bromide, pyridinium, 1-ethyl-, bromide, n-ethylpyridinium bromide, pyridinium, 1-ethyl-, bromide 1:1, pyridineethobromide, ethylpyridiniumbromide, 1-ethyl-pyridiniubromide, n-ethyl pyridinium bromide
PubChem CID 15927
IUPAC Name 1-ethylpyridin-1-ium;bromide
SMILES CC[N+]1=CC=CC=C1.[Br-]

Specifications

Melting Point 117°C to 121°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 188.07
Percent Purity 99%
Chemical Name or Material 1-Ethylpyridinium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-607-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.