Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-2-nitrobenzene, 99%

Catalog No. AC119480500
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1493-27-2 | C6H4FNO2 | 141.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1493-27-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.1
MDL Number MFCD00007048
InChI Key PWKNBLFSJAVFAB-UHFFFAOYSA-N
Synonym 2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro
PubChem CID 73895
IUPAC Name 1-fluoro-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -9°C to 6°C
Color Brown to Yellow
Density 1.3350g/mL
Boiling Point 215°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5307 to 1.5327
Linear Formula FC6H4NO2
Quantity 50 mL
Beilstein 05, 241
Specific Gravity 1.335
Solubility Information Solubility in water: immiscible
Formula Weight 141.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-2-nitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical
EINECSNumber 216-088-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.