Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-3-(trifluoromethoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. F056925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F056925G 25 g
F05695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F056925G Supplier TCI America Supplier No. F056925G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1077-01-6
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.102
MDL Number MFCD00236323
InChI Key AUKDFDQPJWJEDH-UHFFFAOYSA-N
PubChem CID 2777286
IUPAC Name 1-fluoro-3-(trifluoromethoxy)benzene
SMILES C1=CC(=CC(=C1)F)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 105°C
Quantity 25 g
UN Number 1993
Formula Weight 180.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Fluoro-3-(trifluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.