Learn More
1-Fluoro-4-nitrobenzene, 99%
Description
1-Fluoro-4-nitrobenzene used as a component of hair dyes, as pharmaceutical intermediates. It is also used for the preparation of novel soluble aromatic polyimides
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Fluoro-4-nitrobenzene used as a component of hair dyes, as pharmaceutical intermediates. It is also used for the preparation of novel soluble aromatic polyimides
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.

Specifications
Specifications
Melting Point | 22°C to 24°C |
Density | 1.33 |
Boiling Point | 204°C to 206°C |
Flash Point | 83°C (181°F) |
Refractive Index | 1.5315 |
Quantity | 2500 g |
UN Number | UN2811 |
Beilstein | 606923 |
Solubility Information | Insoluble in water. |
Formula Weight | 141.1 |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H331-H335-H373 |
Hazard Statement | GHS H Statement H331-H373-H302-H312-H315-H319 Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 206-502-8 |
RTECSNumber | DA1400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.