Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-4-nitrobenzene, 99%

Catalog No. p-135068
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 350-46-9 | C6H4FNO2 | 141.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-46-9
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.1
MDL Number MFCD00007282
InChI Key WFQDTOYDVUWQMS-UHFFFAOYSA-N
Synonym 4-fluoronitrobenzene, p-fluoronitrobenzene, p-nitrofluorobenzene, benzene, 1-fluoro-4-nitro, 4-nitrofluorobenzene, 1-nitro-4-fluorobenzene, 4-fluoro-1-nitrobenzene, 1-fluoro-4-nitro-benzene, unii-a2m2fh7xhh, p-fluoro nitrobenzene
PubChem CID 9590
IUPAC Name 1-fluoro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 27°C
Color Brown to Yellow
Density 1.3300g/mL
Boiling Point 205°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5302 to 1.5322
Linear Formula FC6H4NO2
Quantity 100 g
Beilstein 05, 241
Specific Gravity 1.33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,xylene and acetone
Formula Weight 141.1
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 1-Fluoro-4-nitrobenzene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
M
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dus
EINECSNumber 206-502-8
RTECSNumber DA1400000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.