Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fmoc-piperidine-4-carboxamidoxime, 97%, Thermo Scientific™

Catalog No. AAH5867203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5867203 1 g
AAH5867206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5867203 Supplier Thermo Scientific Chemicals Supplier No. H5867203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824819-33-1
Molecular Formula C21H23N3O3
Molecular Weight (g/mol) 365.433
MDL Number MFCD22988968
InChI Key ROHLBMBCFADFMX-UHFFFAOYSA-N
Synonym 1-fmoc-piperidine-4-carboxamidoxime
PubChem CID 121235650
IUPAC Name 9H-fluoren-9-ylmethyl 4-[(Z)-N'-hydroxycarbamimidoyl]piperidine-1-carboxylate
SMILES C1CN(CCC1C(=NO)N)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 1 g
Formula Weight 365.43
Percent Purity 97%
Chemical Name or Material 1-Fmoc-piperidine-4-carboxamidoxime
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.