Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Formyl-4-piperidinecarboxylic Acid 95.0+%, TCI America™
SDP

Catalog No. F07491G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 84163-42-8
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.169
MDL Number MFCD03425206
InChI Key IZNTWTMLHCGAJU-UHFFFAOYSA-N
Synonym 1-Formylisonipecotic Acid
PubChem CID 2759232
IUPAC Name 1-formylpiperidine-4-carboxylic acid
SMILES C1CN(CCC1C(=O)O)C=O

Specifications

Melting Point 138°C
Color White
Quantity 1 g
Formula Weight 157.17
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Formyl-4-piperidinecarboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.