Learn More
1-Formylpiperidine, 99%
Description
1-Formylpiperidine reacts with Grignard or alkyllithium reagents gives aldehydes in high yield. It can be used as a polar aprotic solvent, with better hydrocarbon solubility than other amide solvents such as dimethylformamide. In some formylation reaction of alkyllithium compounds it gives higher yields than the DMF.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Formylpiperidine reacts with Grignard or alkyllithium reagents gives aldehydes in high yield. It can be used as a polar aprotic solvent, with better hydrocarbon solubility than other amide solvents such as dimethylformamide. In some formylation reaction of alkyllithium compounds it gives higher yields than the DMF.
Solubility
Freely soluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Specifications
Specifications
Melting Point | -31°C |
Density | 1.019 |
Boiling Point | 222°C |
Flash Point | 102°C (215°F) |
Refractive Index | 1.484 |
Quantity | 1000 g |
Beilstein | 107697 |
Solubility Information | Freely soluble in water. |
Formula Weight | 113.16 |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | H227-H302-H311-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H311-H227-H315-H319-H335 Toxic if swallowed. Toxic in contact with skin. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 219-986-0 |
RTECSNumber | TN0380000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.