Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptanol, 98%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
111-70-6
C7H16O
116.2
MFCD00002986
BBMCTIGTTCKYKF-UHFFFAOYSA-N
1-heptanol,heptyl alcohol,heptanol,n-heptanol,n-heptyl alcohol,enanthic alcohol,gentanol,1-hydroxyheptane,n-heptan-1-ol,enanthyl alcohol
8129
heptan-1-ol
CCCCCCCO
111-70-6
C7H16O
116.2
MFCD00002986
BBMCTIGTTCKYKF-UHFFFAOYSA-N
1-heptanol,heptyl alcohol,heptanol,n-heptanol,n-heptyl alcohol,enanthic alcohol,gentanol,1-hydroxyheptane,n-heptan-1-ol,enanthyl alcohol
8129
heptan-1-ol
CCCCCCCO
Infrared Spectrum Authentic
Color Undesignated
Melting Point -35.0°C
Water 0.5% max.
Specific Gravity 0.8219
Residue after Evaporation 0.01% max.
Packaging Glass bottle
Flash Point 73°C
Quantity 250mL
Density 0.8219g/mL
Acidity 0.5mg KOH/g max.
Refractive Index 1.4230 to 1.4250
Boiling Point 176.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)6OH
Beilstein 01, 414
Merck Index 15, 4696
Solubility Information Solubility in water: 2.85g/L (100°C). Other solubilities: 5.15g/L water (130°C), soluble in ethanol, ether and usual organic, solvents
Formula Weight 116.2
Physical Form Liquid
Percent Purity 98%
Grade Extra Pure
Chemical Name or Material 1-Heptanol
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning