Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptene 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
592-76-7
C7H14
98.19
MFCD00009531
ZGEGCLOFRBLKSE-UHFFFAOYSA-N
1-heptene,heptene,heptylene,n-hept-1-ene,1-n-heptene,unii-o748kj11v7,1-heptene, analytical standard,n-heptene,.alpha.-heptylene,heptene petroleum
11610
hept-1-ene
CCCCCC=C
592-76-7
C7H14
98.19
MFCD00009531
ZGEGCLOFRBLKSE-UHFFFAOYSA-N
1-heptene,heptene,heptylene,n-hept-1-ene,1-n-heptene,unii-o748kj11v7,1-heptene, analytical standard,n-heptene,.alpha.-heptylene,heptene petroleum
11610
hept-1-ene
CCCCCC=C
Specific Gravity 0.697
Infrared Spectrum Authentic
Flash Point −9°C
Refractive Index 1.3995
Packaging Glass bottle
Boiling Point 94.0°C
Melting Point -119.0°C
Density 0.6970g/mL
Color Colorless
Quantity 25mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)4CH=CH2
Beilstein 01, 219
Formula Weight 98.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Heptene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger