Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexadecanol, 98%

Catalog No. p-7081278
Click to view available options
Quantity:
500 g
2500 g

CAS: 36653-82-4 | C16H34O | 242.45 g/mol

1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in alcohol, chloroform, ether

Notes
Keep away from direct sunlight. Incompatible with strong oxidizing agents, strong acids. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
PubChem CID 2682
ChEBI CHEBI:16125
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

Melting Point 47°C to 51°C
Density 0.817 g/mL
Boiling Point 190°C (15 mm)
Flash Point 135°C (275°F)
Assay Percent Range 98%
Quantity 500 g
Beilstein 1748475
Merck Index 14,2028
Solubility Information Soluble in alcohol, chloroform, ether
Formula Weight 242.45
Percent Purity 98%
Chemical Name or Material 1-Hexadecanol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 253-149-0
RTECSNumber MM0225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.