Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexanol, 99%

Catalog No. p-7024058
Click to view available options
Quantity:
500 mL
2500 mL

CAS: 111-27-3 | C6H14O | 102.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.

Solubility
Miscible with ethanol, acetone, chloroform, ether, benzene. Slightly miscible with carbon tetrachloride and water.

Notes
Incompatible with strong acids, acid chlorides, acid anhydrides and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Melting Point °C to 52°C
Density 0.814 g/mL
Boiling Point 157°C
Flash Point 60°C (140°F)
Assay Percent Range 99%
Refractive Index 1.418
Linear Formula CH3(CH2)5OH
Quantity 500 mL
UN Number UN2282
Beilstein 969167
Merck Index 14,4697
Solubility Information Miscible with ethanol,acetone,chloroform,ether,benzene. Slightly miscible with carbon tetrachloride and water.
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 1-Hexanol
Show More Show Less
Hazard Category H226-H302+H312-H319
Hazard Statement GHS H Statement
H226-H302
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P362-P370+P378q-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
EINECSNumber 203-852-3
RTECSNumber MQ4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.