Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexylamine, 99%

Catalog No. aaa15663ak
Click to view available options
Quantity:
250 mL
1000 mL

CAS: 111-26-2 | C6H15N | 101.19 g/mol

1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.

Solubility
Miscible with water, dichloromethane, alcohol and acetone.

Notes
Air sensitive. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-26-2
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008240
InChI Key BMVXCPBXGZKUPN-UHFFFAOYSA-N
Synonym hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
PubChem CID 8102
ChEBI CHEBI:5712
IUPAC Name hexan-1-amine
SMILES CCCCCCN

Specifications

Melting Point -19°C
Density 0.773
pH 11.6
Boiling Point 129°C to 131°C
Flash Point 27°C (80°F)
Odor Amine-like
Refractive Index 1.418
Linear Formula CH3(CH2)5NH2
Quantity 250 mL
UN Number UN2734
Beilstein 1731298
Sensitivity Air Sensitive
Solubility Information Miscible with water,dichloromethane,alcohol and acetone.
Formula Weight 101.19
Percent Purity 99%
Chemical Name or Material 1-Hexylamine
Show More Show Less
Hazard Category H226-H301+H311-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 203-851-8
RTECSNumber MQ4540000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.