Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexylboronic acid pinacol ester, 98%

Catalog No. AAH2783106
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 86308-26-1 | C12H25BO2 | 212.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86308-26-1
Molecular Formula C12H25BO2
Molecular Weight (g/mol) 212.14
MDL Number MFCD08457665
InChI Key LDPMCSWZEGKWLC-UHFFFAOYSA-N
Synonym hexylboronic acid pinacol ester, 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 1,3,2-dioxaborolane, 2-hexyl-4,4,5,5-tetramethyl
PubChem CID 10921795
IUPAC Name 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCCCCC

Specifications

Density 1.167
Flash Point 84°C (183°F)
Refractive Index 1.426
Quantity 5 g
Formula Weight 212.14
Percent Purity 98%
Chemical Name or Material 1-Hexylboronic acid pinacol ester
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.