Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1'-Hydroxy-2'-acetonaphthone 98.0+%, TCI America™
SDP

Catalog No. H0935100G
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 711-79-5
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00003963
InChI Key JBGJVMVWYWUVOW-UHFFFAOYSA-N
Synonym 1'-hydroxy-2'-acetonaphthone, 2-acetyl-1-naphthol, 1-hydroxy-2-acetylnaphthalene, 2-acetyl-1-hydroxynaphthalene, 1-hydroxy-2-acetonaphthone, ethanone, 1-1-hydroxy-2-naphthalenyl, 1-1-hydroxynaphthalen-2-yl ethanone, 1-1-hydroxy-2-naphthalenyl ethanone, 2'-acetonaphthone, 1'-hydroxy, 1-hydroxy-2-naphthyl methyl ketone
PubChem CID 69733
IUPAC Name 1-(1-hydroxynaphthalen-2-yl)ethan-1-one
SMILES CC(=O)C1=CC=C2C=CC=CC2=C1O

Specifications

Melting Point 99°C
Quantity 100 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1′-Hydroxy-2′-acetonaphthone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.