Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxybenzotriazole, 98%, Thermo Scientific™

Catalog No. AC169160050
Click to view available options
:
5g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2592-95-2
CAS Min % 97.5
CAS Max % 100.0
Molecular Formula C6H8N4O
MDL Number MFCD0006569
Quantity 5g
Fieser 03,156; 05,342; 06,288
Synonym hobt, n-hydroxybenzotriazole, 1h-1,2,3-benzotriazol-1-ol, 1h-benzotriazol-1-ol, 1h-benzo d 1,2,3 triazol-1-ol, 1h-benzotriazole, 1-hydroxy, benzazimidol, benzazimidol hydrate, 1-hydroxy-1h-benzotriazole, unii-a2t929dmg4
InChI Key URQAMACHHWVNRD-UHFFFAOYSA-N
SMILES N.ON1N=NC2=CC=CC=C12
IUPAC Name 1H-1,2,3-benzotriazol-1-ol amine
Molecular Weight (g/mol) 152.16
PubChem CID 75771
Formula Weight 135.12
Chemical Name or Material 1-Hydroxybenzotriazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 219-989-7
RTECSNumber DM1288000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.