Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Indanone, 99+%

Catalog No. AAA1105622
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

CAS: 83-33-0 | C9H8O | 132.162 g/mol

1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.

Solubility
Soluble in water (6.5 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-33-0
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00003785
InChI Key QNXSIUBBGPHDDE-UHFFFAOYSA-N
Synonym 1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
PubChem CID 6735
ChEBI CHEBI:17404
IUPAC Name 2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=CC=CC=C21

Specifications

Melting Point 38°C to 42°C
Density 1.103
Boiling Point 243°C to 245°C
Flash Point 113°C (235°F)
Quantity 100 g
Beilstein 507957
Solubility Information Soluble in water (6.5g/L at 20°C).
Formula Weight 132.16
Percent Purity ≥99%
Chemical Name or Material 1-Indanone
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-470-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.