Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodoheptane, 98%, stab. with copper

Click to view available options
Quantity:
50g
250g
4282-40-0
C7H15I
226.101
MFCD00001104
LMHCYRULPLGEEZ-UHFFFAOYSA-N
heptyl iodide,n-heptyl iodide,heptane, 1-iodo,iodoheptane,n-heptyliodide,iodo heptane,1-iodanylheptane,1-iodomeptane,1-iodo heptane,1-iodo-heptane
20274
1-iodoheptane
CCCCCCCI
4282-40-0
C7H15I
226.101
MFCD00001104
LMHCYRULPLGEEZ-UHFFFAOYSA-N
heptyl iodide,n-heptyl iodide,heptane, 1-iodo,iodoheptane,n-heptyliodide,iodo heptane,1-iodanylheptane,1-iodomeptane,1-iodo heptane,1-iodo-heptane
20274
1-iodoheptane
CCCCCCCI
Applications
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
Boiling Point 203°C to 205°C
Quantity 50g
Refractive Index 1.49
Sensitivity Light sensitive
Density 1.379
Flash Point 78°C (174°F)
Melting Point -48°C
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 226.1
Percent Purity 98%
Chemical Name or Material 1-Iodoheptane, stabilized with copper
Show More Show Less

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 224-285-8

RTECSNumber : MJ1050000

TSCA : Yes

Recommended Storage : Ambient temperatures