Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodohexadecane 95%, Thermo Scientific™

Catalog No. AC412431000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412431000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412431000 Supplier Thermo Scientific Chemicals Supplier No. 412431000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-77-4
Molecular Formula C16H33I
Molecular Weight (g/mol) 352.344
MDL Number MFCD00001089
InChI Key KMWHQYDMBYABKL-UHFFFAOYSA-N
Synonym cetyl iodide, hexadecyl iodide, hexadecane, 1-iodo, hexadecane, iodo, unii-jo95x48m8o, 1-iodo-hexadecan, iodohexadecane, cetyljodid, 1-iodo-hexadecane, n-hexadecyl iodide
PubChem CID 11007
IUPAC Name 1-iodohexadecane
SMILES CCCCCCCCCCCCCCCCI

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Golden or Pink to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4800 to 1.4830 (20°C, 589nm)
Linear Formula CH3(CH2)15I
Quantity 100 g
Beilstein 01, 172
Merck Index 09, 1984
Formula Weight 352.33
Percent Purity ≥94%
Physical Form Liquid After Melting
Chemical Name or Material 1-Iodohexadecane, 95%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.