Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodooctadecane 98.0+%, TCI America™
SDP

Catalog No. I0065100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0065100G 100 g
I006525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0065100G Supplier TCI America Supplier No. I0065100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 629-93-6
Molecular Formula C18H37I
Molecular Weight (g/mol) 380.398
MDL Number MFCD00001090
InChI Key ZNJOCVLVYVOUGB-UHFFFAOYSA-N
Synonym Octadecyl Iodide, Stearyl Iodide
PubChem CID 12402
IUPAC Name 1-iodooctadecane
SMILES CCCCCCCCCCCCCCCCCCI

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 197°C
Quantity 100 g
Formula Weight 380.40
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 1-Iodooctadecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.