Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodooctane (stabilized with Copper chip) 97.0+%, TCI America™
SDP

Catalog No. I0321100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0321100G 100 g
I032125G 25 g
I0321500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0321100G Supplier TCI America Supplier No. I0321100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 629-27-6
Molecular Formula C8H17I
Molecular Weight (g/mol) 240.128
MDL Number MFCD00001105
InChI Key UWLHSHAHTBJTBA-UHFFFAOYSA-N
Synonym n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
PubChem CID 12380
IUPAC Name 1-iodooctane
SMILES CCCCCCCCI

Specifications

Color Red-Yellow
Boiling Point 226°C
Quantity 100 g
Formula Weight 240.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodooctane (stabilized with Copper chip)
RTECSNumber RG9700100
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.